missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Hydroxyxanthen-9-one 98.0+%, TCI America™
$58.06 - $177.51
Chemical Identifiers
CAS | 3722-51-8 |
---|---|
Molecular Formula | C13H8O3 |
Molecular Weight (g/mol) | 212.204 |
MDL Number | MFCD00604748 |
InChI Key | XCJHDJAODLKGLG-UHFFFAOYSA-N |
Synonym | sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone |
PubChem CID | 5376013 |
IUPAC Name | 3-hydroxyxanthen-9-one |
SMILES | C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O |
Chemical Identifiers
3722-51-8 | |
212.204 | |
XCJHDJAODLKGLG-UHFFFAOYSA-N | |
5376013 | |
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O |
C13H8O3 | |
MFCD00604748 | |
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone | |
3-hydroxyxanthen-9-one |
Specifications
3722-51-8 | |
White-Yellow | |
MFCD00604748 | |
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone | |
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O | |
212.204 | |
212.20 | |
Crystalline Powder |
243°C | |
C13H8O3 | |
1 g | |
XCJHDJAODLKGLG-UHFFFAOYSA-N | |
3-hydroxyxanthen-9-one | |
5376013 | |
≥98.0% (HPLC,T) | |
3-Hydroxyxanthen-9-one |
Safety and Handling
TSCA : No