Please login to your online account to display your discounted pricing

3-Hydroxypyridine-N-oxide 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  153580250

 View more versions of this product

Catalog No. AC153580250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 111.1
Quantity 25g
Color Light yellow to beige
RTECS Number UU7719100
TSCA TSCA
Melting Point 189° to 192°C
EINECS Number 229-545-4
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 3-Pyridinol N-oxide
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Health Hazard 1 Warning
CAS 6602-28-4
Chemical Name or Material 3-Hydroxypyridine-N-oxide
Name Note 99%
MDL Number MFCD00006200
Molecular Formula C5H5NO2

Specifications

Chemical Name or Material 3-Hydroxypyridine-N-oxide
Name Note 99%
CAS 6602-28-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H5NO2
Formula Weight 111.1
MDL Number MFCD00006200
Synonym 3-Pyridinol N-oxide
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents