Learn More
3-Hydroxypyridine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 121851000
Specifications
3-Hydroxypyridine | |
Plastic bottle | |
180.0°C (22.0 mmHg) | |
123.0°C to 130.0°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
MFCD00006378 | |
01,486 | |
Solubility in water: 33g/L water (20°C). | |
C1=CC(=CN=C1)O | |
95.1 | |
95.1 | |
98% |
Authentic | |
0.2% max. | |
Beige to Brown | |
100g | |
109-00-2 | |
100.0 | |
C5H5NO | |
21, 46 | |
3-hydoxypyridine, 3-hydroxypyridine, 3-oxopyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-pyridyl alcohol, beta-hydroxypyridine, m-hydroxypyridine, unii-4kbe4p5b6s | |
GRFNBEZIAWKNCO-UHFFFAOYSA-N | |
pyridin-3-ol | |
7971 | |
Crystalline Powder |
Chemical Identifiers
109-00-2 | |
95.1 | |
GRFNBEZIAWKNCO-UHFFFAOYSA-N | |
7971 | |
C1=CC(=CN=C1)O |
C5H5NO | |
MFCD00006378 | |
3-hydoxypyridine, 3-hydroxypyridine, 3-oxopyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-pyridyl alcohol, beta-hydroxypyridine, m-hydroxypyridine, unii-4kbe4p5b6s | |
pyridin-3-ol |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-637-4
RTECSNumber : UU7701400
TSCA : TSCA