missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxypyridine-4-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H6346506

 View more versions of this product

Catalog No. AAH6346506



Description

Description

Applications
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3-Hydroxypyridine-4-carboxylic acid
10128-71-9
MFCD00234165
3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid
C1=CN=CC(=C1C(=O)O)O
139.11
139.11
98%
5g
C6H5NO3
383677
JEHGATQUCUYHJL-UHFFFAOYSA-N
3-hydroxypyridine-4-carboxylic acid
459503
Solid

Chemical Identifiers

10128-71-9
139.11
JEHGATQUCUYHJL-UHFFFAOYSA-N
459503
C1=CN=CC(=C1C(=O)O)O
C6H5NO3
MFCD00234165
3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid
3-hydroxypyridine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents