Please login to your online account to display your discounted pricing

3-Hydroxypiperidine hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hydroxypiperidine hydrochloride
CAS 64051-79-2
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C5H11NO·ClH
Formula Weight 137.61
Synonym 3-Piperidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC174320050

  SDS
thermo fisher scientific
AC174320050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC174320250

  SDS
thermo fisher scientific
AC174320250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC174321000

  SDS
thermo fisher scientific
AC174321000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.61
Physical Form Powder
Quantity 5g
Color White
Melting Point 195° to 200°C
CAS Max % 100.0
Infrared Spectrum Authentic
Elemental Analysis 42.97 to 44.28% C, 9.90 to 10.33% N
Synonym 3-Piperidinol hydrochloride
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C5H11NO·ClH
Assay Percent Range 99%
Chemical Name or Material 3-Hydroxypiperidine hydrochloride
CAS 64051-79-2
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.