missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxyphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC359410100

 View more versions of this product

Catalog No. AC359410100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Hydroxyphenylboronic acid
Glass bottle
212.0°C
87199-18-6
100.0
C6H7BO3
3-boronophenol, 3-hydroxybenzeneboronic acid, 3-hydroxyphenyboronic acid, 3-hydroxyphenyl boranediol, 3-hydroxyphenyl boronic acid, 3-hydroxyphenylboronicacid, acmc-209qip, boronic acid, 3-hydroxyphenyl, m-hydroxyphenylboronic acid, pubchem1728
B(C1=CC(=CC=C1)O)(O)O
137.93
137.93
97%
Authentic
Pink to Tan
10g
96.0
96% min. (HPLC)
HOC6H4B(OH)2
WFWQWTPAPNEOFE-UHFFFAOYSA-N
(3-hydroxyphenyl)boronic acid
2734359
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

87199-18-6
137.93
3-boronophenol, 3-hydroxybenzeneboronic acid, 3-hydroxyphenyboronic acid, 3-hydroxyphenyl boranediol, 3-hydroxyphenyl boronic acid, 3-hydroxyphenylboronicacid, acmc-209qip, boronic acid, 3-hydroxyphenyl, m-hydroxyphenylboronic acid, pubchem1728
(3-hydroxyphenyl)boronic acid
C6H7BO3
WFWQWTPAPNEOFE-UHFFFAOYSA-N
2734359
B(C1=CC(=CC=C1)O)(O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents