missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxyphenylacetylene, 97%

Manufacturer:  Alfa Aesar™ L1152303

 View more versions of this product

Catalog No. AAL1152303



Description

Description

Applications
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

3-Hydroxyphenylacetylene
1g
C8H6O
2076613
AODMJIOEGCBUQL-UHFFFAOYSA-N
3-ethynylphenol
139144
97%
231°C
10401-11-3
MFCD00078347
3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci, 3-hydroxyphenylacetylene
C#CC1=CC(=CC=C1)O
118.135
118.14

Chemical Identifiers

10401-11-3
118.135
AODMJIOEGCBUQL-UHFFFAOYSA-N
139144
C#CC1=CC(=CC=C1)O
C8H6O
MFCD00078347
3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci, 3-hydroxyphenylacetylene
3-ethynylphenol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280g-P305+P351+P338-P310
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

Documents