Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Hydroxymethyl)benzeneboronic acid, 98%

$66.61 - $176.30

Chemical Identifiers

Chemical Name or Material 3-(Hydroxymethyl)benzeneboronic acid
CAS 87199-15-3
Molecular Formula C7H9BO3
Formula Weight 151.96
MDL Number MFCD01317846
Beilstein 8832019
Synonym 3-(Hydroxymethyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1519303

  SDS
alfa aesar™
L1519303
1g Each for $66.61

AAL1519306

  SDS
alfa aesar™
L1519306
5g Each for $176.30
Description & Specifications

Specifications

Formula Weight 151.96
Chemical Name or Material 3-(Hydroxymethyl)benzeneboronic acid
CAS 87199-15-3
Synonym 3-(Hydroxymethyl)phenylboronic acid
Melting Point 93° to 98°C
MDL Number MFCD01317846
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H9BO3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Beilstein 8832019
Health Hazard 1 Warning
Assay 98%

Applications
Reactant used for copper-mediated trifluoromethylation, copper-catalyzed transformations from arylboronic acids in water, Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate. Reactant involved in the synthesis of biologically active molecules including Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions, HIV protease inhibitors with antiviral activity against drug-resistant viruses, Pyrrole derivatives for use as PDE4B inhibitors.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.