missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Hydroxymethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1519306

 View more versions of this product

Catalog No. AAL1519306



Description

Description

Reactant used for copper-mediated trifluoromethylation, copper-catalyzed transformations from arylboronic acids in water, Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate. Reactant involved in the synthesis of biologically active molecules including Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions, HIV protease inhibitors with antiviral activity against drug-resistant viruses, Pyrrole derivatives for use as PDE4B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for copper-mediated trifluoromethylation, copper-catalyzed transformations from arylboronic acids in water, Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate. Reactant involved in the synthesis of biologically active molecules including Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions, HIV protease inhibitors with antiviral activity against drug-resistant viruses, Pyrrole derivatives for use as PDE4B inhibitors.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3-(Hydroxymethyl)benzeneboronic acid
93°C to 98°C
MFCD01317846
8832019
Soluble in water.
OCC1=CC=CC(=C1)B(O)O
151.96
151.96
87199-15-3
C7H9BO3
5 g
3-hydroxymethyl phenylboronic acid, 3-hydroxymethylphenylboronic acid, 3-hydroxymethyl phenyl boronic acid, 3-hydroxymethyl benzeneboronic acid, 3-boronobenzyl alcohol, 3-hydroxymethylphenyl boronic acid, 3-hydroxymethyl phenylboronicacid, 3-hydroxymethyl phenyl boranediol, 3-boronophenyl methanol
HGTDLKXUWVKLQX-UHFFFAOYSA-N
[3-(hydroxymethyl)phenyl]boronic acid
2734662
98%

Chemical Identifiers

87199-15-3
151.96
HGTDLKXUWVKLQX-UHFFFAOYSA-N
2734662
OCC1=CC=CC(=C1)B(O)O
C7H9BO3
MFCD01317846
3-hydroxymethyl phenylboronic acid, 3-hydroxymethylphenylboronic acid, 3-hydroxymethyl phenyl boronic acid, 3-hydroxymethyl benzeneboronic acid, 3-boronobenzyl alcohol, 3-hydroxymethylphenyl boronic acid, 3-hydroxymethyl phenylboronicacid, 3-hydroxymethyl phenyl boranediol, 3-boronophenyl methanol
[3-(hydroxymethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only