missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxymethyl-4-nitroindole, 97%

Manufacturer:  Alfa Aesar™ H63120MD

 View more versions of this product

Catalog No. AAH63120MD



Specifications

Specifications

3-Hydroxymethyl-4-nitroindole
250mg
C9H8N2O3
4-nitro-1h-indol-3-yl methanol, 4-nitro-1h-indol-3-yl-methanol
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)CO
192.174
192.17
97%
Undesignated
124549-51-5
MFCD04973980
GIZXLVHVDBXUPQ-UHFFFAOYSA-N
(4-nitro-1H-indol-3-yl)methanol
14432522
Solid

Chemical Identifiers

124549-51-5
192.174
GIZXLVHVDBXUPQ-UHFFFAOYSA-N
14432522
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)CO
C9H8N2O3
MFCD04973980
4-nitro-1h-indol-3-yl methanol, 4-nitro-1h-indol-3-yl-methanol
(4-nitro-1H-indol-3-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents