missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Hydroxymethyl)-4-iodophenol, 95%, Maybridge

Chemical Identifiers

CAS 915707-73-2
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C7H7IO2
Formula Weight 250.03
MDL Number MFCD09065032
Chemical Name or Material 3-(Hydroxymethyl)-4-iodophenol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO07996CB SDS Fisher Scientific Company
MO07996CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

915707-73-2
95%
250.03
3-(Hydroxymethyl)-4-iodophenol
95.0
C7H7IO2
MFCD09065032
95%

Specifications

250.03
95.0
915707-73-2
MFCD09065032
3-(Hydroxymethyl)-4-iodophenol
250mg
Amber Glass Bottle
C7H7IO2
95%
95%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning