Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxymethyl-1-adamantanol, 96%

Chemical Identifiers

Chemical Name or Material 3-Hydroxymethyl-1-adamantanol
CAS 38584-37-1
Formula Weight 182.26
MDL Number MFCD03105096
Synonym 1-Hydroxy-3-adamantanemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6672103

  SDS
alfa aesar™
H6672103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6672106

  SDS
alfa aesar™
H6672106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66721MD

  SDS
alfa aesar™
H66721MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 158° to 162°C
Formula Weight 182.26
Chemical Name or Material 3-Hydroxymethyl-1-adamantanol
MDL Number MFCD03105096
Assay 96%
CAS 38584-37-1
Physical Form Powder
Quantity 1g
Color White to pale brown
Synonym 1-Hydroxy-3-adamantanemethanol