missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxyisoquinoline, TCI America™

$96.25 - $333.06

Chemical Identifiers

CAS 7651-81-2
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00075524
InChI Key GYPOFOQUZZUVQL-UHFFFAOYSA-N
Synonym 3-hydroxyisoquinoline, isoquinolin-3-ol, isoquinolin-3 2h-one, 3-isoquinolinol, 2,3-dihydroisoquinolin-3-one, 3 2h-isoquinolinone, 2h-3-isoquinolone, zlchem 163, 3-hydroxy-isoquinoline, 3-isoquinolinol 9ci
PubChem CID 2736554
IUPAC Name 2H-isoquinolin-3-one
SMILES C1=CC2=CC(=O)NC=C2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14511G View Documents TCI America
H14511G
1g Each for $96.25
 
H14515G View Documents TCI America
H14515G
5g Each for $333.06
 
Specifications

Chemical Identifiers

7651-81-2
145.161
GYPOFOQUZZUVQL-UHFFFAOYSA-N
2736554
C1=CC2=CC(=O)NC=C2C=C1
C9H7NO
MFCD00075524
3-hydroxyisoquinoline, isoquinolin-3-ol, isoquinolin-3 2h-one, 3-isoquinolinol, 2,3-dihydroisoquinolin-3-one, 3 2h-isoquinolinone, 2h-3-isoquinolone, zlchem 163, 3-hydroxy-isoquinoline, 3-isoquinolinol 9ci
2H-isoquinolin-3-one

Specifications

White-Yellow
1g
C9H7NO
3-hydroxyisoquinoline, isoquinolin-3-ol, isoquinolin-3 2h-one, 3-isoquinolinol, 2,3-dihydroisoquinolin-3-one, 3 2h-isoquinolinone, 2h-3-isoquinolone, zlchem 163, 3-hydroxy-isoquinoline, 3-isoquinolinol 9ci
C1=CC2=CC(=O)NC=C2C=C1
145.161
145.16
≥98.0% (GC)
194°C
7651-81-2
MFCD00075524
GYPOFOQUZZUVQL-UHFFFAOYSA-N
2H-isoquinolin-3-one
2736554
Crystal-Powder at 20°C
3-Hydroxyisoquinoline
Safety and Handling

Safety and Handling

TSCA : No