Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Hydroxyflavanone, 98%

$168.76 - $626.81

Chemical Identifiers

Chemical Name or Material 3'-Hydroxyflavanone
CAS 1621-55-2
Molecular Formula C15H12O3
Formula Weight 240.25
MDL Number MFCD00017738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2510903

  SDS
alfa aesar™
B2510903
1g Each for $168.76

AAB2510906

  SDS
alfa aesar™
B2510906
5g Each for $626.81
Description & Specifications

Specifications

Formula Weight 240.25
CAS 1621-55-2
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3'-Hydroxyflavanone
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 139° to 141°C
MDL Number MFCD00017738
Molecular Formula C15H12O3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.