Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxydiphenylamine, 98%


Quantity 100g

Manufacturer: alfa aesar™  L0367822

 View more versions of this product

Catalog No. AAL0367822

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 185.23
Chemical Name or Material 3-Hydroxydiphenylamine
CAS 101-18-8
Beilstein 2092957
Synonym 3-Anilinophenol
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 98%
Molecular Formula C12H11NO
MDL Number MFCD00002262
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 340°C
Melting Point 78° to 82°C
Quantity 100g
Health Hazard 1 Danger

Specifications

Chemical Name or Material 3-Hydroxydiphenylamine
CAS 101-18-8
Molecular Formula C12H11NO
Formula Weight 185.23
MDL Number MFCD00002262
Beilstein 2092957
Synonym 3-Anilinophenol
Accessories

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents