missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxybutyric acid, tech.

Manufacturer:  Alfa Aesar™ 4335406

 View more versions of this product

Catalog No. AA4335406



Specifications

Specifications

3-Hydroxybutyric acid
>112°C (234°F)
Hygroscopic
Colorless
300-85-6
MFCD00004546
3 hba
WHBMMWSBFZVSSR-UHFFFAOYSA-N
3-hydroxybutanoic acid
441
104.1
Technical
1.13g/mL
1.4445
130°C (12mmHg)
5g
C4H8O3
14,4816
Soluble in water, alcohol, ether
CC(CC(=O)O)O
104.105
CHEBI:20067
Liquid

Chemical Identifiers

300-85-6
104.105
WHBMMWSBFZVSSR-UHFFFAOYSA-N
441
3-hydroxybutanoic acid
C4H8O3
MFCD00004546
3 hba
CHEBI:20067
CC(CC(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents