missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxybutyric acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121050050

 View more versions of this product

Catalog No. AC121050050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Hydroxybutyric acid
112°C
1.4420 to 1.4440
118.0°C to 120.0°C (2.0 mmHg)
300-85-6
100.0
C4H8O3
MFCD00004546
14,178
3 hba, 3 hydroxybutyrate, 3-hydroxybuttersaeure, 3-hydroxybutyric acid, beta-hydroxy-n-butyric acid, beta-hydroxybuttersaeure, beta-hydroxybutyric acid, butanoic acid, 3-hydroxy, butyric acid, 3-hydroxy, dl-beta-hydroxybutyric acid
WHBMMWSBFZVSSR-UHFFFAOYSA-N
3-hydroxybutanoic acid
441
104.11
Technical
1.1260g/mL
Glass bottle
1.126
5g
94.0
95&percent;
CH3CH(OH)CH2CO2H
03, 307
15, 4855
Solubility in water: >10% in water. Other solubilities: soluble in alcohol,ether
CC(CC(=O)O)O
104.11
CHEBI:20067
95%

Chemical Identifiers

300-85-6
104.11
WHBMMWSBFZVSSR-UHFFFAOYSA-N
441
3-hydroxybutanoic acid
C4H8O3
MFCD00004546
3 hba, 3 hydroxybutyrate, 3-hydroxybuttersaeure, 3-hydroxybutyric acid, beta-hydroxy-n-butyric acid, beta-hydroxybuttersaeure, beta-hydroxybutyric acid, butanoic acid, 3-hydroxy, butyric acid, 3-hydroxy, dl-beta-hydroxybutyric acid
CHEBI:20067
CC(CC(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advic

GHS Signal Word: Warning

EINECSNumber : 206-099-9

SDS Documents