missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxybenzylhydrazine dihydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 229030050

 View more versions of this product

Catalog No. AC229030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Hydroxybenzylhydrazine dihydrochloride
Glass bottle
138°C
98%
97.5
98%
HOC6H4CH2NHNH2·2HCl
3-hydrazinomethyl phenol dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride, 3-hydrazinomethyl-phenol dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydroxybenzylhydrazine dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, alpha-hydrazino-m-cresol dihydrochloride, m-hydroxybenzylhydrazine dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride
C1=CC(=CC(=C1)O)CNN.Cl.Cl
211.09
211.09
98%
Authentic
White to Beige
5g
81012-99-9
100.0
C7H10N22HCl
MFCD00044695
ONOJPUDFIOEGCX-UHFFFAOYSA-N
3-(hydrazinylmethyl)phenol;dihydrochloride
2724402
Adhering Powder

Chemical Identifiers

81012-99-9
211.09
ONOJPUDFIOEGCX-UHFFFAOYSA-N
2724402
C1=CC(=CC(=C1)O)CNN.Cl.Cl
C7H10N22HCl
MFCD00044695
3-hydrazinomethyl phenol dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride, 3-hydrazinomethyl-phenol dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydroxybenzylhydrazine dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, alpha-hydrazino-m-cresol dihydrochloride, m-hydroxybenzylhydrazine dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride
3-(hydrazinylmethyl)phenol;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 279-659-3

TSCA : TSCA

SDS Documents