Learn More
3-Hydroxybenzylhydrazine dihydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 229030010
Specifications
3-Hydroxybenzylhydrazine dihydrochloride | |
Glass bottle | |
138°C | |
98% | |
97.5 | |
98% | |
HOC6H4CH2NHNH2·2HCl | |
3-hydrazinomethyl phenol dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride, 3-hydrazinomethyl-phenol dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydroxybenzylhydrazine dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, alpha-hydrazino-m-cresol dihydrochloride, m-hydroxybenzylhydrazine dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride | |
C1=CC(=CC(=C1)O)CNN.Cl.Cl | |
211.09 | |
211.09 | |
98% |
Authentic | |
White to Beige | |
1g | |
81012-99-9 | |
100.0 | |
C7H10N2O·2HCl | |
MFCD00044695 | |
ONOJPUDFIOEGCX-UHFFFAOYSA-N | |
3-(hydrazinylmethyl)phenol;dihydrochloride | |
2724402 | |
Adhering Powder |
Chemical Identifiers
81012-99-9 | |
211.09 | |
ONOJPUDFIOEGCX-UHFFFAOYSA-N | |
2724402 | |
C1=CC(=CC(=C1)O)CNN.Cl.Cl |
C7H10N2O·2HCl | |
MFCD00044695 | |
3-hydrazinomethyl phenol dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride, 3-hydrazinomethyl-phenol dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydroxybenzylhydrazine dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, alpha-hydrazino-m-cresol dihydrochloride, m-hydroxybenzylhydrazine dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride | |
3-(hydrazinylmethyl)phenol;dihydrochloride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 279-659-3
TSCA : TSCA