missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 81012-99-9
Molecular Formula C7H12Cl2N2O
Molecular Weight (g/mol) 211.09
MDL Number MFCD00044695
InChI Key ONOJPUDFIOEGCX-UHFFFAOYSA-N
Synonym 3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
PubChem CID 2724402
IUPAC Name 3-(hydrazinylmethyl)phenol;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229030010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
229030010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC229030050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
229030050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

81012-99-9
211.09
ONOJPUDFIOEGCX-UHFFFAOYSA-N
2724402
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
C7H12Cl2N2O
MFCD00044695
3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
3-(hydrazinylmethyl)phenol;dihydrochloride

Specifications

81012-99-9
100.0
98%
C7H12Cl2N2O
MFCD00044695
3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1
211.09
211.09
3-Hydroxybenzylhydrazine dihydrochloride, 98%
97.5
Authentic
Glass bottle
HOC6H4CH2NHNH2·2HCl
1g
ONOJPUDFIOEGCX-UHFFFAOYSA-N
3-(hydrazinylmethyl)phenol;dihydrochloride
2724402
98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 279-659-3

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only