missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxybenzyl Alcohol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC121025000

 View more versions of this product

Catalog No. AC121025000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Hydroxybenzyl alcohol
Glass bottle
67°C to 71°C
620-24-6
100.0
C7H8O2
MFCD00004643
3-hydroxybenzenemethanol, 3-hydroxybenzyl alcohol, 3-hydroxybenzylalcohol, 3-hydroxymethyl phenol, 3-hydroxymethyl-phenol, 3-methylolphenol, 3-oh-benzyl-alcohol, benzenemethanol, 3-hydroxy, benzyl alcohol, m-hydroxy, m-hydroxybenzyl alcohol
OKVJCVWFVRATSG-UHFFFAOYSA-N
3-(hydroxymethyl)phenol
102
124.14
97%
Authentic
Pink or Beige to Brown
500g
96.0
96% min. (GC)
HOC6H4CH2OH
06, 896
Solubility in water: soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol and ether
C1=CC(=CC(=C1)O)CO
124.14
CHEBI:17069
Crystalline Powder

Chemical Identifiers

620-24-6
124.14
OKVJCVWFVRATSG-UHFFFAOYSA-N
102
3-(hydroxymethyl)phenol
C7H8O2
MFCD00004643
3-hydroxybenzenemethanol, 3-hydroxybenzyl alcohol, 3-hydroxybenzylalcohol, 3-hydroxymethyl phenol, 3-hydroxymethyl-phenol, 3-methylolphenol, 3-oh-benzyl-alcohol, benzenemethanol, 3-hydroxy, benzyl alcohol, m-hydroxy, m-hydroxybenzyl alcohol
CHEBI:17069
C1=CC(=CC(=C1)O)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

EINECSNumber : 210-633-6

SDS Documents