missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Hydroxybenzyl Alcohol, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 121020050
Specifications
3-Hydroxybenzyl alcohol | |
Glass bottle | |
67°C to 71°C | |
620-24-6 | |
100.0 | |
C7H8O2 | |
MFCD00004643 | |
3-hydroxybenzenemethanol, 3-hydroxybenzyl alcohol, 3-hydroxybenzylalcohol, 3-hydroxymethyl phenol, 3-hydroxymethyl-phenol, 3-methylolphenol, 3-oh-benzyl-alcohol, benzenemethanol, 3-hydroxy, benzyl alcohol, m-hydroxy, m-hydroxybenzyl alcohol | |
OKVJCVWFVRATSG-UHFFFAOYSA-N | |
3-(hydroxymethyl)phenol | |
102 | |
124.14 | |
97% |
Authentic | |
Beige to Brown or Pink | |
5g | |
96.0 | |
96% min. (GC) | |
HOC6H4CH2OH | |
06, 896 | |
Solubility in water: soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol and ether | |
C1=CC(=CC(=C1)O)CO | |
124.14 | |
CHEBI:17069 | |
Crystalline Powder |
Chemical Identifiers
620-24-6 | |
124.14 | |
OKVJCVWFVRATSG-UHFFFAOYSA-N | |
102 | |
3-(hydroxymethyl)phenol |
C7H8O2 | |
MFCD00004643 | |
3-hydroxybenzenemethanol, 3-hydroxybenzyl alcohol, 3-hydroxybenzylalcohol, 3-hydroxymethyl phenol, 3-hydroxymethyl-phenol, 3-methylolphenol, 3-oh-benzyl-alcohol, benzenemethanol, 3-hydroxy, benzyl alcohol, m-hydroxy, m-hydroxybenzyl alcohol | |
CHEBI:17069 | |
C1=CC(=CC(=C1)O)CO |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
EINECSNumber : 210-633-6