Please login to your online account to display your discounted pricing

3-Hydroxybenzophenone, 99%, ACROS Organics™

$221.08 - $221.08

Chemical Identifiers

Chemical Name or Material 3-Hydroxybenzophenone
Name Note 99%
CAS 13020-57-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H10O2
Linear Formula HOC6H4COC6H5
Formula Weight 198.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205190050

  SDS
thermo fisher scientific
205190050
Glass bottle 5g Each for $221.08
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 198.22
Physical Form Crystalline Powder
Quantity 5g
Color Light beige to brown-gray
Beilstein 08, 157
TSCA TSCA
EINECS Number 235-879-1
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00002297
CAS 13020-57-0
Name Note 99%
Linear Formula HOC6H4COC6H5
Chemical Name or Material 3-Hydroxybenzophenone
Molecular Formula C13H10O2
Assay Percent Range 99%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.