missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxybenzonitrile, 99%

$104.84 - $347.85

Chemical Identifiers

CAS 873-62-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002252
InChI Key SGHBRHKBCLLVCI-UHFFFAOYSA-N
Synonym 3-cyano phenol, 3-cyano-phenol, 3-cyanophenol, 3-hydroxy benzonitrile, 3-hydroxy-benzonitrile, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, m-cyanophenol, m-hydroxybenzonitrile, pubchem13113
PubChem CID 13394
IUPAC Name 3-hydroxybenzonitrile
SMILES C1=CC(=CC(=C1)O)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2170206
View Documents
Alfa Aesar™
B2170206
5g
Each for $104.84
 
AAB2170214
View Documents
Alfa Aesar™
B2170214
25g
Each for $347.85
 
Description

Description

Applications
3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity in vivo.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

873-62-1
119.123
SGHBRHKBCLLVCI-UHFFFAOYSA-N
13394
C1=CC(=CC(=C1)O)C#N
C7H5NO
MFCD00002252
3-cyano phenol, 3-cyano-phenol, 3-cyanophenol, 3-hydroxy benzonitrile, 3-hydroxy-benzonitrile, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, m-cyanophenol, m-hydroxybenzonitrile, pubchem13113
3-hydroxybenzonitrile

Specifications

78°C to 81°C
873-62-1
MFCD00002252
Slightly soluble in water.
C1=CC(=CC(=C1)O)C#N
119.123
119.12
3-Hydroxybenzonitrile
5g
C7H5NO
3-cyano phenol, 3-cyano-phenol, 3-cyanophenol, 3-hydroxy benzonitrile, 3-hydroxy-benzonitrile, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, m-cyanophenol, m-hydroxybenzonitrile, pubchem13113
SGHBRHKBCLLVCI-UHFFFAOYSA-N
3-hydroxybenzonitrile
13394
99%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 212-845-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS