missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxybenzoic acid, 99%

$64.65 - $253.99

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Formula Weight 138.12
MDL Number MFCD00002506
Beilstein 508160
Chemical Name or Material 3-Hydroxybenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AAA1362822 SDS Alfa Aesar™
A1362822
100g Each for $64.65
AAA1362836 SDS Alfa Aesar™
A1362836
500g Each for $253.99
Specifications

Description

Applications
3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

99-06-9
138.12
508160
C7H6O3
MFCD00002506
3-Hydroxybenzoic acid

Specifications

220°C (428°F)
201° to 203°C
99-06-9
138.12
508160
99%
Odorless
100g
C7H6O3
MFCD00002506
3-Hydroxybenzoic acid

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DH1924980

TSCA : TSCA