missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxyaspartic Acid, TCI America™

$145.14 - $400.28

Chemical Identifiers

CAS 71653-06-0
Molecular Formula C4H7NO5
Molecular Weight (g/mol) 149.102
MDL Number MFCD00128126
InChI Key YYLQUHNPNCGKJQ-UHFFFAOYSA-N
Synonym 2-Amino-3-hydroxysuccinic Acid
PubChem CID 5425
ChEBI CHEBI:83981
IUPAC Name 2-amino-3-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09475G View Documents TCI America
H09475G
5g Each for $400.28
 
H09471G View Documents TCI America
H09471G
1g Each for $145.14
 
Specifications

Chemical Identifiers

71653-06-0
149.102
YYLQUHNPNCGKJQ-UHFFFAOYSA-N
5425
2-amino-3-hydroxybutanedioic acid
C4H7NO5
MFCD00128126
2-Amino-3-hydroxysuccinic Acid
CHEBI:83981
C(C(C(=O)O)O)(C(=O)O)N

Specifications

White
71653-06-0
MFCD00128126
YYLQUHNPNCGKJQ-UHFFFAOYSA-N
2-amino-3-hydroxybutanedioic acid
5425
149.10
≥98.0% (T,N)
5g
C4H7NO5
2-Amino-3-hydroxysuccinic Acid
C(C(C(=O)O)O)(C(=O)O)N
149.102
CHEBI:83981
Crystal-Powder at 20°C
3-Hydroxyaspartic Acid
Safety and Handling

Safety and Handling

TSCA : No