missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxyadamantane-1-carboxylic acid, 97%, Maybridge

Chemical Identifiers

CAS 42711-75-1
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00193929
InChI Key CJJMAWPEZKYJAP-UHFFFAOYSA-N
Synonym 1-carboxy-3-adamantanol
PubChem CID 2736518
IUPAC Name 3-hydroxyadamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12757EA SDS View Documents Fisher Scientific Company
BTB12757EA
10g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

42711-75-1
196.246
CJJMAWPEZKYJAP-UHFFFAOYSA-N
2736518
C1C2CC3(CC1CC(C2)(C3)O)C(=O)O
C11H16O3
MFCD00193929
1-carboxy-3-adamantanol
3-hydroxyadamantane-1-carboxylic acid

Specifications

Amber Glass Bottle
42711-75-1
100.0
C11H16O3
1-carboxy-3-adamantanol
C1C2CC3(CC1CC(C2)(C3)O)C(=O)O
196.246
196.24
3-Hydroxyadamantane-1-carboxylic acid
10g
96.5
97%
MFCD00193929
CJJMAWPEZKYJAP-UHFFFAOYSA-N
3-hydroxyadamantane-1-carboxylic acid
2736518
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.