Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxyadamantane-1-acetic acid, 97%

$95.54 - $95.54

Chemical Identifiers

Chemical Name or Material 3-Hydroxyadamantane-1-acetic acid
CAS 17768-36-4
Molecular Formula C12H18O3
Formula Weight 210.27
MDL Number MFCD00167797
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2764503

  SDS
alfa aesar™
H2764503
1g Each for $95.54
Description & Specifications

Specifications

Formula Weight 210.27
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C12H18O3
Melting Point 125° to 129°C
MDL Number MFCD00167797
CAS 17768-36-4
Chemical Name or Material 3-Hydroxyadamantane-1-acetic acid
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.