missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-6-methylpyridine 99%, ACROS Organics™

Chemical Identifiers

CAS 1121-78-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H7NO
Linear Formula (C5H4N)CH2OH
Formula Weight 109.13
MDL Number MFCD00006339
Synonym 6-Methyl-3-pyridinol
Chemical Name or Material 3-Hydroxy-6-methylpyridine, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC148110250 SDS Fisher Scientific Acros
148110250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1121-78-4
100.0
C6H7NO
109.13
6-Methyl-3-pyridinol
≥98.5%
98.5
99%
(C5H4N)CH2OH
MFCD00006339
3-Hydroxy-6-methylpyridine, 99%

Specifications

109.13
25g
21, III, 480
100.0
Glass bottle
6-Methyl-3-pyridinol
MFCD00006339
1121-78-4
3-Hydroxy-6-methylpyridine, 99%
Crystalline Powder
Beige to light tan
168° to 171°C
98.5
Authentic
(C5H4N)CH2OH
99%
C6H7NO
≥98.5%

Safety and Handling

EINECSNumber : 214-337-8

RTECSNumber : UU7714900

TSCA : TSCA