Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Hydroxy-6-methylflavone, 97%

$265.03 - $265.03

Chemical Identifiers

Chemical Name or Material 3'-Hydroxy-6-methylflavone
CAS 468060-75-5
Molecular Formula C16H12O3
Formula Weight 252.28g/mol
MDL Number MFCD07778551
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3062414

 
alfa aesar™
H30624-14
25g Each for $265.03
Description & Specifications

Specifications

Melting Point 203° to 206°C
Quantity 25g
Chemical Name or Material 3'-Hydroxy-6-methylflavone
CAS 468060-75-5
Assay 97%
Molecular Formula C16H12O3
Formula Weight 252.28g/mol
MDL Number MFCD07778551
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.