missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-5-methylisoxazole, 97%

Manufacturer:  Alfa Aesar™ 4445006

 View more versions of this product

Catalog No. AA4445006



Specifications

Specifications

3-Hydroxy-5-methylisoxazole
10004-44-1
MFCD00144468
3-hydroxy-5-methylisoxazole
KGVPNLBXJKTABS-UHFFFAOYSA-N
5-methyl-1,2-oxazol-3-one
24781
99.09
97%
5g
C4H5NO2
14,4856
Soluble in alcohol, acetone, THF, chloroform
CC1=CC(=O)NO1
99.089
CHEBI:5827
Powder

Chemical Identifiers

10004-44-1
99.089
KGVPNLBXJKTABS-UHFFFAOYSA-N
24781
5-methyl-1,2-oxazol-3-one
C4H5NO2
MFCD00144468
3-hydroxy-5-methylisoxazole
CHEBI:5827
CC1=CC(=O)NO1
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 233-000-6

RTECSNumber : NY2932000

TSCA : No

Recommended Storage : Ambient temperatures

Documents