missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-5-methylisoxazole, 97%

Manufacturer:  Alfa Aesar™ 4445022

 View more versions of this product

Catalog No. AA4445022



Specifications

Specifications

3-Hydroxy-5-methylisoxazole
10004-44-1
MFCD00144468
hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
KGVPNLBXJKTABS-UHFFFAOYSA-N
5-methyl-1,2-oxazol-3-one
24781
99.09
97%
100g
C4H5NO2
14,4856
Soluble in alcohol, acetone, THF, chloroform
CC1=CC(=O)NO1
99.089
CHEBI:5827
Powder

Chemical Identifiers

10004-44-1
99.089
KGVPNLBXJKTABS-UHFFFAOYSA-N
24781
5-methyl-1,2-oxazol-3-one
C4H5NO2
MFCD00144468
hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
CHEBI:5827
CC1=CC(=O)NO1
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

RTECSNumber : NY2932000

SDS Documents