missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 121670250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Hydroxy-4-nitrobenzoic acid | |
619-14-7 | |
100.0 | |
98% | |
C7H5NO5 | |
MFCD00007110 | |
10, 146 | |
Solubility in water: insoluble | |
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O | |
183.12 | |
183.12 |
98% | |
97.5 | |
Authentic | |
Plastic bottle | |
HOC6H3(NO2)CO2H | |
25g | |
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid | |
XLDLRRGZWIEEHT-UHFFFAOYSA-N | |
3-hydroxy-4-nitrobenzoic acid | |
69265 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 210-580-9
TSCA : TSCA
RUO â Research Use Only