missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-4-nitrobenzaldehyde, TCI America™

$42.78 - $140.56

Chemical Identifiers

CAS 704-13-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007109
InChI Key AUBBVPIQUDFRQI-UHFFFAOYSA-N
Synonym benzaldehyde, 3-hydroxy-4-nitro, 3-hydroxy-4-nitro-benzaldehyde, 4-nitro-3-hydroxybenzaldehyde, 3-formyl-6-nitrophenol, acmc-1bm06, ksc495a6r, 3-hydroxy-4-nitro benzaldehyde, 3-hydroxy-4-nitrobenzaldehyde
PubChem CID 69712
IUPAC Name 3-hydroxy-4-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13341G View Documents TCI America
H13341G
1g Each for $42.78
 
H13345G View Documents TCI America
H13345G
5g Each for $140.56
 
Specifications

Chemical Identifiers

704-13-2
167.12
AUBBVPIQUDFRQI-UHFFFAOYSA-N
69712
C1=CC(=C(C=C1C=O)O)[N+](=O)[O-]
C7H5NO4
MFCD00007109
benzaldehyde, 3-hydroxy-4-nitro, 3-hydroxy-4-nitro-benzaldehyde, 4-nitro-3-hydroxybenzaldehyde, 3-formyl-6-nitrophenol, acmc-1bm06, ksc495a6r, 3-hydroxy-4-nitro benzaldehyde, 3-hydroxy-4-nitrobenzaldehyde
3-hydroxy-4-nitrobenzaldehyde

Specifications

Undesignated
1g
C7H5NO4
benzaldehyde, 3-hydroxy-4-nitro, 3-hydroxy-4-nitro-benzaldehyde, 4-nitro-3-hydroxybenzaldehyde, 3-formyl-6-nitrophenol, acmc-1bm06, ksc495a6r, 3-hydroxy-4-nitro benzaldehyde, 3-hydroxy-4-nitrobenzaldehyde
C1=CC(=C(C=C1C=O)O)[N+](=O)[O-]
167.12
167.12
≥97.0% (GC,T)
131°C
704-13-2
MFCD00007109
AUBBVPIQUDFRQI-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzaldehyde
69712
Crystal-Powder at 20°C
3-Hydroxy-4-nitrobenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No