missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-4-methoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 269410-23-3
Molecular Formula C13H19BO4
Molecular Weight (g/mol) 250.10
MDL Number MFCD09027077
InChI Key WINSAMQIAMEZIK-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid pinacol ester, 3-hydroxy-4-methoxyphenylboronic acid, pinacol ester, amtb953, phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 17750251
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445060010
SDS View Documents
Thermo Fisher Scientific
AC445060010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445060050
SDS View Documents
Thermo Fisher Scientific
AC445060050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

269410-23-3
250.10
WINSAMQIAMEZIK-UHFFFAOYSA-N
17750251
COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1
C13H19BO4
MFCD09027077
2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid pinacol ester, 3-hydroxy-4-methoxyphenylboronic acid, pinacol ester, amtb953, phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

Glass Bottle
1g
96% min. (GC)
MFCD09027077
WINSAMQIAMEZIK-UHFFFAOYSA-N
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
17750251
97%
101.0°C to 102.0°C
269410-23-3
C13H19BO4
2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid pinacol ester, 3-hydroxy-4-methoxyphenylboronic acid, pinacol ester, amtb953, phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1
250.10
250.1
3-Hydroxy-4-methoxyphenylboronic acid pinacol ester
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS