Learn More
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1048203
Description
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.
Solubility
Soluble in ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications
3-Hydroxy-4-methoxycinnamic acid | |
537-73-5 | |
C10H10O4 | |
1 g | |
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid | |
QURCVMIEKCOAJU-HWKANZROSA-N | |
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid | |
736186 | |
194.19 |
predominantly trans | |
∼231°C (decomposition) | |
MFCD00004391 | |
2212760 | |
Soluble in ethanol and methanol. | |
COC1=C(C=C(C=C1)C=CC(=O)O)O | |
194.186 | |
CHEBI:27794 | |
≥98% |
Chemical Identifiers
537-73-5 | |
194.186 | |
QURCVMIEKCOAJU-HWKANZROSA-N | |
736186 | |
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid |
C10H10O4 | |
MFCD00004391 | |
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid | |
CHEBI:27794 | |
COC1=C(C=C(C=C1)C=CC(=O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-676-0
RTECSNumber : UD3365400
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only