Please login to your online account to display your discounted pricing

3-Hydroxy-4-methoxycinnamic Acid, Predominantly trans 97%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146890010

 View more versions of this product

Catalog No. AC146890010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder or Flakes
Formula Weight 194.19
Quantity 1g
Color Off-white to beige
EINECS Number 208-676-0
TSCA TSCA
RTECS Number UD3365400
Melting Point 228° to 233°C
Beilstein 10, 437
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H10O4
Linear Formula HOC6H3(OCH3)CH=CHCO2H
MDL Number MFCD00004391
CAS 537-73-5
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid
Name Note 97%
CAS 537-73-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O4
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Formula Weight 194.19
MDL Number MFCD00004391
Accessories

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents