Please login to your online account to display your discounted pricing

3-Hydroxy-4-methoxycinnamic Acid, Predominantly trans 97%, Acros Organics

$45.33 - $45.33

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid
Name Note 97%
CAS 537-73-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O4
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Formula Weight 194.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146890050

  SDS
thermo fisher scientific
146890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC146890010

  SDS
thermo fisher scientific
146890010
Glass bottle 1g Each for $45.33
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder or Flakes
Formula Weight 194.19
Quantity 5g
Color Off-white to beige
Beilstein 10, 437
RTECS Number UD3365400
TSCA TSCA
Melting Point 228° to 233°C
EINECS Number 208-676-0
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid
MDL Number MFCD00004391
CAS 537-73-5
Molecular Formula C10H10O4
Name Note 97%
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Assay Percent Range 97%
Safety and Handling

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.