Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-4-methoxybenzyl alcohol, 98%

$55.45 - $194.38

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-4-methoxybenzyl alcohol
CAS 4383-06-6
Molecular Formula C8H10O3
Formula Weight 154.17
MDL Number MFCD00004644
Beilstein 1867860
Synonym Isovanillyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1478609

  SDS
alfa aesar™
A1478609
10g Each for $55.45

AAA1478618

  SDS
alfa aesar™
A1478618
50g Each for $194.38
Description & Specifications

Specifications

Formula Weight 154.17
Chemical Name or Material 3-Hydroxy-4-methoxybenzyl alcohol
MDL Number MFCD00004644
Molecular Formula C8H10O3
Melting Point 135° to 137°C
Quantity 10g
Beilstein 1867860
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Synonym Isovanillyl alcohol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 4383-06-6
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.