missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-4-methoxybenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ A1478618

 View more versions of this product

Catalog No. AAA1478618



Specifications

Specifications

3-Hydroxy-4-methoxybenzyl alcohol
50g
C10H12O
1867860
ZGYBYYJGIKPBFD-UHFFFAOYNA-N
2-methyl-1-phenylprop-2-en-1-ol
138211
98%
135°C to 137°C
4383-08-8
MFCD00004644
2-methyl-1-phenyl-2-propen-1-ol, acmc-1bn9z, alpha-isopropenylbenzenemethanol, 1-phenyl-2-methyl-2-propen-1-ol, 2,6-dichloro-9-methyl-10h-purine
CC(=C)C(C1=CC=CC=C1)O
148.21
154.17

Chemical Identifiers

4383-08-8
148.21
ZGYBYYJGIKPBFD-UHFFFAOYNA-N
138211
CC(=C)C(C1=CC=CC=C1)O
C10H12O
MFCD00004644
2-methyl-1-phenyl-2-propen-1-ol, acmc-1bn9z, alpha-isopropenylbenzenemethanol, 1-phenyl-2-methyl-2-propen-1-ol, 2,6-dichloro-9-methyl-10h-purine
2-methyl-1-phenylprop-2-en-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents