missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-4-methoxybenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ A1478609

 View more versions of this product

Catalog No. AAA1478609



Specifications

Specifications

3-Hydroxy-4-methoxybenzyl alcohol
10g
C8H10O3
1867860
WHKRHBLAJFYZKF-UHFFFAOYSA-N
5-(hydroxymethyl)-2-methoxyphenol
78089
98%
135°C to 137°C
4383-06-6
MFCD00004644
3-hydroxy-4-methoxybenzyl alcohol, 5-hydroxymethyl-2-methoxyphenol, isovanillyl alcohol, 3-hydroxy-4-methoxybenzylalcohol, benzenemethanol, 3-hydroxy-4-methoxy, 4-methoxy-3-hydroxy benzyl alcohol, pubchem15050, ksc495i5l, 3-hydroxy-4-methoxy-benzyl alcohol, 3-hydroxy-4-methoxy benzyl alcohol
COC1=C(C=C(C=C1)CO)O
154.17
154.17

Chemical Identifiers

4383-06-6
154.17
WHKRHBLAJFYZKF-UHFFFAOYSA-N
78089
COC1=C(C=C(C=C1)CO)O
C8H10O3
MFCD00004644
3-hydroxy-4-methoxybenzyl alcohol, 5-hydroxymethyl-2-methoxyphenol, isovanillyl alcohol, 3-hydroxy-4-methoxybenzylalcohol, benzenemethanol, 3-hydroxy-4-methoxy, 4-methoxy-3-hydroxy benzyl alcohol, pubchem15050, ksc495i5l, 3-hydroxy-4-methoxy-benzyl alcohol, 3-hydroxy-4-methoxy benzyl alcohol
5-(hydroxymethyl)-2-methoxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents