Please login to your online account to display your discounted pricing

3-Hydroxy-4-methoxybenzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-4-methoxybenzoic acid
Name Note 99%
CAS 645-08-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8O4
Linear Formula HOC6H3(OCH3)CO2H
Formula Weight 168.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205030050

  SDS
fisher scientific acros
AC205030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205030250

  SDS
thermo fisher scientific
205030250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 168.15
Quantity 5g
Color Beige to gray-brown
EINECS Number 211-430-5
TSCA TSCA
RTECS Number BZ4850000
Melting Point 245° to 250°C
Beilstein 10, 393
CAS Max % 100.0
Chemical Name or Material 3-Hydroxy-4-methoxybenzoic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H8O4
Linear Formula HOC6H3(OCH3)CO2H
MDL Number MFCD00002507
CAS 645-08-9
Synonym Isovanillic acid
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.