Please login to your online account to display your discounted pricing

3-Hydroxy-4-iodobenzaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10430DA

 View more versions of this product

Catalog No. BTB10430DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 248.01
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 3-Hydroxy-4-iodobenzaldehyde
Assay Percent Range 97%
Molecular Formula C7H5IO2
CAS 135242-71-6

Specifications

Chemical Name or Material 3-Hydroxy-4-iodobenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5IO2
Formula Weight 248.01