Please login to your online account to display your discounted pricing

3-Hydroxy-4-iodobenzaldehyde, 97%, Maybridge

$152.71 - $1,120.28

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-4-iodobenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5IO2
Formula Weight 248.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10430DA

  SDS
fisher scientific company
BTB10430DA
Amber Glass Bottle 1g Each for $152.71

BTB10430EA

  SDS
fisher scientific company
BTB10430EA
Amber Glass Bottle 10g Each for $1,120.28
Description & Specifications

Specifications

Formula Weight 248.01
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 3-Hydroxy-4-iodobenzaldehyde
Assay Percent Range 97%
Molecular Formula C7H5IO2
CAS 135242-71-6