missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-4,5-dimethoxybenzaldehyde, TCI America™

$132.92 - $397.23

Chemical Identifiers

CAS 29865-90-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00010442
InChI Key NVLTWXMZECWWPC-UHFFFAOYSA-N
Synonym 5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde
PubChem CID 141513
IUPAC Name 3-hydroxy-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H11791G View Documents TCI America
H11791G
1g Each for $132.92
 
H11795G View Documents TCI America
H11795G
5g Each for $397.23
 
Specifications

Chemical Identifiers

29865-90-5
182.175
NVLTWXMZECWWPC-UHFFFAOYSA-N
141513
COC1=CC(=CC(=C1OC)O)C=O
C9H10O4
MFCD00010442
5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde
3-hydroxy-4,5-dimethoxybenzaldehyde

Specifications

180°C
66°C
29865-90-5
MFCD00010442
NVLTWXMZECWWPC-UHFFFAOYSA-N
3-hydroxy-4,5-dimethoxybenzaldehyde
141513
Crystal-Powder at 20°C
3-Hydroxy-4,5-dimethoxybenzaldehyde
Undesignated
1g
C9H10O4
5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde
COC1=CC(=CC(=C1OC)O)C=O
182.175
182.18
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No