missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-4,4′-dimethyl-2,2′-bipyridyl, TCI America™

$381.95 - $1,332.25

Chemical Identifiers

CAS 81998-07-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD28339195
InChI Key AWUFFAVGWFOSNV-KHPPLWFESA-N
Synonym 4,4′C-Dimethyl-(2,2′C-Bipyridin)-3-ol
PubChem CID 118798227
IUPAC Name (2Z)-4-methyl-2-(4-methyl-1H-pyridin-2-ylidene)pyridin-3-one
SMILES CC1=CC(=C2C(=O)C(=CC=N2)C)NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15561G View Documents TCI America
H15561G
1g Each for $1,332.25
 
H1556200MG View Documents TCI America
H1556200MG
200mg Each for $381.95
 
Specifications

Chemical Identifiers

81998-07-4
200.241
AWUFFAVGWFOSNV-KHPPLWFESA-N
118798227
CC1=CC(=C2C(=O)C(=CC=N2)C)NC=C1
C12H12N2O
MFCD28339195
4,4′C-Dimethyl-(2,2′C-Bipyridin)-3-ol
(2Z)-4-methyl-2-(4-methyl-1H-pyridin-2-ylidene)pyridin-3-one

Specifications

Yellow
1g
C12H12N2O
4,4′C-Dimethyl-(2,2′C-Bipyridin)-3-ol
CC1=CC(=C2C(=O)C(=CC=N2)C)NC=C1
200.241
200.24
≥98.0% (GC,T)
110°C
81998-07-4
MFCD28339195
AWUFFAVGWFOSNV-KHPPLWFESA-N
(2Z)-4-methyl-2-(4-methyl-1H-pyridin-2-ylidene)pyridin-3-one
118798227
Crystal-Powder at 20°C
3-Hydroxy-4,4′-dimethyl-2,2′-bipyridyl
Safety and Handling

Safety and Handling

TSCA : No