Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-4-(2,2,2-trimethylacetamido)pyridine, 98%

$100.51 - $344.80

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-4-(2,2,2-trimethylacetamido)pyridine
CAS 169205-93-0
Molecular Formula C10H14N2O2
Formula Weight 194.24
MDL Number MFCD07776841
Synonym N-(3-Hydroxypyridin-4-yl)pivalamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5003503

 
alfa aesar™
H5003503
1g Each for $344.80

AAH50035MD

 
alfa aesar™
H50035MD
250mg Each for $100.51
Description & Specifications

Specifications

Formula Weight 194.24
Chemical Name or Material 3-Hydroxy-4-(2,2,2-trimethylacetamido)pyridine
Assay 98%
Molecular Formula C10H14N2O2
MDL Number MFCD07776841
CAS 169205-93-0
Synonym N-(3-Hydroxypyridin-4-yl)pivalamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.