missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-3-methylvaleric Acid, TCI America™

$223.06 - $716.55

Chemical Identifiers

CAS 150-96-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00059657
InChI Key KEGHVPSZIWXTPY-UHFFFAOYSA-N
Synonym 3-Hydroxy-3-methylpentanoic Acid
PubChem CID 538288
IUPAC Name 3-hydroxy-3-methylpentanoic acid
SMILES CCC(C)(CC(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H06265G View Documents TCI America
H06265G
5g Each for $223.06
 
H062625G View Documents TCI America
H062625G
25g Each for $716.55
 
Specifications

Chemical Identifiers

150-96-9
132.159
KEGHVPSZIWXTPY-UHFFFAOYSA-N
538288
CCC(C)(CC(=O)O)O
C6H12O3
MFCD00059657
3-Hydroxy-3-methylpentanoic Acid
3-hydroxy-3-methylpentanoic acid

Specifications

144°C
5g
C6H12O3
3-Hydroxy-3-methylpentanoic Acid
CCC(C)(CC(=O)O)O
132.159
132.16
≥95.0% (GC,T)
Yellow
150-96-9
MFCD00059657
KEGHVPSZIWXTPY-UHFFFAOYSA-N
3-hydroxy-3-methylpentanoic acid
538288
Clear Liquid at 20°C
3-Hydroxy-3-methylvaleric Acid
Safety and Handling

Safety and Handling

TSCA : No