Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-3-methyl-2-butanone oxime, 98%

$64.34 - $218.49

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone oxime
CAS 7431-25-6
Molecular Formula C5H11NO2
Formula Weight 117.15
MDL Number MFCD00059656
Beilstein 1747660
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1062406

  SDS
alfa aesar™
L1062406
5g Each for $64.34

AAL1062414

  SDS
alfa aesar™
L1062414
25g Each for $218.49
Description & Specifications

Specifications

Formula Weight 117.15
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone oxime
MDL Number MFCD00059656
Molecular Formula C5H11NO2
Assay 98%
Beilstein 1747660
Quantity 5g
Melting Point 84° to 86°C
CAS 7431-25-6