missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-3-methyl-2-butanone oxime, 98%

Manufacturer:  Alfa Aesar™ L1062406

 View more versions of this product

Catalog No. AAL1062406



Specifications

Specifications

3-Hydroxy-3-methyl-2-butanone oxime
5g
C5H11NO2
1747660
QKLLBCGVADVPKD-XQRVVYSFSA-N
(3Z)-3-hydroxyimino-2-methylbutan-2-ol
5385758
98%
84°C to 86°C
7431-25-6
MFCD00059656
3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
CC(=NO)C(C)(C)O
117.148
117.15

Chemical Identifiers

7431-25-6
117.148
QKLLBCGVADVPKD-XQRVVYSFSA-N
5385758
CC(=NO)C(C)(C)O
C5H11NO2
MFCD00059656
3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
(3Z)-3-hydroxyimino-2-methylbutan-2-ol
Documents